GRAB-EEZ® Cleanroom Wipe Dispenser
Catalogue # 77007-992
Fournisseur: High-Tech Conversions
Spécifications
- Description:GRAB-EEZ® cleanroom wipe dispenser
- Material:Stainless steel
- Cat. no.:77007-992
Spécifications
À propos de cet article
The GRAB-EEZ® cleanroom wipe dispenser, now made from durable stainless steel, is a cost saving solution to wasteful wipe use.
- Designed to eliminate wasteful use
- Wall or table mountable
- Accommodates unique double C-fold GRAB-EEZ® cleanroom wipes
- Suitable for ISO 5 (Class 100) or higher
This easy-to-use wipe dispenser is wall and table-top mountable, and is designed to eliminate wasteful wipe use, cross contamination, and countertop clutter.