Virtu-Clean® Universal Cleaning Pads
Catalogue # 77007-852
Fournisseur: High-Tech Conversions
Spécifications
- Description:Virtu-Clean® Universal cleaning pad
- Packed:20 pads/bag, 12 bags/master bag
- Cat. no.:77007-852
Spécifications
À propos de cet article
Not all cleaning pads are equal. Absorbency rates, release rates, coverage, and linting can vary greatly, significantly impacting the finished product.
- Single-use application to reduce risk of cross contamination
- Embossed design reduces friction for less strain on workers
- Channels efficiently pick up and retain dirt in pad
- No Quat binding
- Compatible with chlorine bleach, hydrogen peroxide, and common disinfectants
- 300 to 360 sq. ft. cleaning coverage
- Absorbs 9X its weight in liquid
Virtu-Clean™ Universal Cleaning Pads are specially engineered for critical environments, providing maximum coverage, cleaning, and convenience. These disposable, single-use mop pads check all the right boxes – they hold more liquid, release that liquid at a controlled rate over a larger surface area, and reduce friction for more effort-free cleaning.