Bee-Write™ ESD Cleanroom Papers | Avantor
Bee-Write™ ESD Cleanroom Papers
Bee-Write™ ESD Cleanroom Papers
Certificates
Bee-Write™ ESD Cleanroom Papers
3 Options Available Below
Product Details & Documents
Bee-Write™ ESD Cleanroom Paper is cleanroom packaged (vacuum packed, double-bagged) and processed in a class 100 ISO 5 controlled environment.
  • Low Chloride and other ionic contamination; ultra-low ionic contamination
  • Formulated without Magnesium Silicate
  • Excellent IPA resistance
  • Compatible to most inks in writing without smearing
  • Ultra-low particle and fiber generation
  • Excellent heat resistance material for laser printing and photocopiers
  • Compatible with laser and inkjet printers
  • Latex-free
  • ROHS compliant
Because Bee-Write™ Cleanroom Paper is ESD safe, it is also suitable for use in a variety of electronics assembly cleanrooms. It is compatible with most pens as well as laser and inkjet printers.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Dimensions
    Packaging
    Availability
    Price
  • ESD cleanroom paper
    Blue
    8.5×11"
    250 sheets/ream
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2,500
    Case of 2,500 - $325.31
    $325.31
    Case of 2,500
       $325.31
    Supplier Number: BW-CRP-8511-BLUE
  • ESD cleanroom paper
    Green
    8.5×11"
    250 sheets/ream
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2,500
    Case of 2,500 - $325.31
    $325.31
    Case of 2,500
       $325.31
    Supplier Number: BW-CRP-8511-GREEN
  • ESD cleanroom paper
    White
    8.5×11"
    250 sheets/ream
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2,500
    Case of 2,500 - $325.31
    $325.31
    Case of 2,500
       $325.31
    Supplier Number: BW-CRP-8511-WHITE