Code Corporation Barcode Reader Accessories
About this item
Accessories available for the barcode readers.
Use the 6′ (176701) or 8' (176700) Coiled RS232 Affinity® Cable CRA-C511 with Code's cabled barcode readers.
Model 176702 Code power supply is an international power supply with adapter clips for the US, EU, UK and AU (barrel plug, 12 V/3.3 A).
Model 176703 Code power supply is an international power supply with USB connection.
Model 176856 - Use this stand to scan hands-free or simply to hold your scanner. It features a flexible gooseneck design that adjusts easily to different sized scanners.
5 Options Available Below
Product Details & Documents
Accessories available for the barcode readers.
Use the 6′ (176701) or 8' (176700) Coiled RS232 Affinity® Cable CRA-C511 with Code's cabled barcode readers.
Model 176702 Code power supply is an international power supply with adapter clips for the US, EU, UK and AU (barrel plug, 12 V/3.3 A).
Model 176703 Code power supply is an international power supply with USB connection.
Model 176856 - Use this stand to scan hands-free or simply to hold your scanner. It features a flexible gooseneck design that adjusts easily to different sized scanners.
Product Family Options
Product Information
- DésignationFor ModelAvailabilityPrice
- Supplier Number: 176701
- Supplier Number: 176700
- Power supply, black, 1.5×3×2.5", 19.7' cable lengthCR2700 Quad bay battery charger (176520)Log in for availabilityEach$76.53Supplier Number: 176702
- Power supply, black, 1.5×3×2.5", 19.7' cable lengthCR2700 Quad bay battery charger (176520)Log in for availabilityEach$76.53Supplier Number: 176703
- Scanner stand, black, 9×4.25×6"CR1500 (176510), CR1500 kit (176511), CR1100 (176697), CR1100 kit (176698), CR950 (176836)Log in for availabilityEach$87.45Supplier Number: 176856