Code Corporation Code Reader™ 5200 Barcode Scanner | Avantor
Code Corporation Code Reader™ 5200 Barcode Scanner
Supplier:   Brady Worldwide
Code Corporation Code Reader™ 5200 Barcode Scanner
Supplier:  Brady Worldwide
Certificates
Code Corporation Code Reader™ 5200 Barcode Scanner
Code Corporation Code Reader™ 5200 Barcode Scanner
2 Options Available Below
Product Details & Documents
Seamlessly connect your operations with the CR5200. It reads barcodes in any orientation, plus offers hands-free operation to expedite workflows.
  • Rotational tolerance ±180°
  • Field of view 51° horizontal by 39.4° vertical
  • CMOS 1.2 megapixel sensor
  • Dark gray color
  • Cabled scanner connectivity (6' cable length)
  • No battery
Get high-speed, omnidirectional reading of 1D and 2D barcode symbologies. Quickly read barcodes on mobile device screens. Compact and adjustable for any reading direction. Provides user feedback with LED and audible tone. Features extended read ranges.
Model 176696 features age verification when scanning ID cards.
Delivery information: Includes (1) CR5200; (1) USB cable.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 77282-158
  • 77282-160
  • Interfaces:
  • RS232; USB 2.0 (generic HID, HID keyboard, virtual COM port) [reader/scanner unit]
  • RS232; USB 2.0 (generic HID, HID keyboard, virtual COM port) [reader/scanner unit]
  • Sensor type:
  • CMOS 1.2 MP
  • CMOS 1.2 MP
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • CR5200 Fixed barcode scanner
    3W×3.5D×5.5"H
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 928,15 $
    928,15 $
    Prix promotionnel pour les nouveaux clients
    Each
       928,15 $
    Supplier Number: 176695
    Spécifications:
    More
    Specifications
    Cat. No.
    77282-158
    Interfaces
    RS232; USB 2.0 (generic HID, HID keyboard, virtual COM port) [reader/scanner unit]
    Sensor type
    CMOS 1.2 MP
  • CR5200 Fixed barcode scanner with age verification
    3W×3.5D×5.5"H
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 146,88 $
    1 146,88 $
    Prix promotionnel pour les nouveaux clients
    Each
       1 146,88 $
    Supplier Number: 176696
    Spécifications:
    More
    Specifications
    Cat. No.
    77282-160
    Interfaces
    RS232; USB 2.0 (generic HID, HID keyboard, virtual COM port) [reader/scanner unit]
    Sensor type
    CMOS 1.2 MP