Aggressive Adhesive Multi-Purpose Nylon Labels
Aggressive Adhesive Multi-Purpose Nylon Labels
Aggressive Adhesive Multi-Purpose Nylon Labels
Aggressive Adhesive Multi-Purpose Nylon Labels
Use these highly-pliable labels for tightly-curved surfaces, like wires and cables. Bulk label rolls are ideal for continuous printing, reducing the frequency of label changeouts. Wraptor™ A6200 bulk label roll adapter required to utilize bulk label rolls.
  • Permanent acrylic adhesive
  • Matte finish
  • Autoclavable, cryogenic storage, halogen-free material
  • Abrasion-resistant, chemical-resistant, humidity-resistant, low temperature-resistant, UV-resistant, and weather-resistant
  • Service temperature: −40 to 194 °F
  • UL approved, CSA accepted
Labels conform to small-gauge wires and cables, and are recommended for indoor use.
Use with Wraptor™ A6200 bulk label roll adapter and Wraptor™ A6200 wrap printer applicator and Wraptor™ A6200 bulk label roll adapter (required). A ribbon is required to print this material; use part number A62-R4300.
Product Family Options
  • Désignation
    Matière
    Pour l'utilisation avec
    Width
    Hauteur
    Packaging
  • Adhesive nylon wire labels, 22 - 6 AWG
    Nylon cloth
    Wraptor™ A6200
    0.5"
    0.75"
    5200/roll
    Roll of 1
    Roll of 1 - $1,177.89
    $1,177.89
    Roll of 1
       $1,177.89
     : 170991
  • Adhesive nylon wire labels, 16 - 6 AWG
    Nylon cloth
    Wraptor™ A6200
    0.5"
    1"
    4000/roll
    Roll of 1
    Roll of 1 - $969.38
    $969.38
    Roll of 1
       $969.38
     : 170992
  • Adhesive nylon wire labels, 22 - 6 AWG
    Nylon cloth
    Wraptor™ A6200
    1"
    0.75"
    5200/roll
    Roll of 1
    Roll of 1 - $1,298.19
    $1,298.19
    Roll of 1
       $1,298.19
     : 170993
  • Adhesive nylon wire labels, 16 - 6 AWG
    Nylon cloth
    Wraptor™ A6200
    1"
    1"
    4000/roll
    Roll of 1
    Roll of 1 - $1,173.56
    $1,173.56
    Roll of 1
       $1,173.56
     : 170994