High-Adhesion Self-Laminating Wire and Cable Labels
About this item
Use high-adhesive, self-laminating labels for tough conditions like high-humidity, small diameter wires, and newer wire jacketing materials. Bulk label rolls are ideal for continuous printing, reducing the frequency of label changeouts. Wraptor™ A6200 bulk label roll adapter required to utilize bulk label rolls.
- Permanent acrylic adhesive
- Matte finish
- Self-laminating, self-extinguishing material
- Abrasion-resistant, chemical-resistant, humidity-resistant, oil-resistant, water-resistant, and weather-resistant
- Service temperature: −40 to 158 °F
Dependable, long-term adhesion to small diameter wires. Labels will stay adhered in even the most humid environments. Strong adhesion to a variety of wire jacketing materials, including Teflon™, silicone, braided and traditional PVC. Labels feature a white printable area and a clear overlaminating area that protects the print. Labels are self-extinguishing and resist chemicals, humidity, oil, and water.
Use with Wraptor™ A6200 bulk label roll adapter and Wraptor™ A6200 wrap printer applicator and Wraptor™ A6200 bulk label roll adapter (required). A ribbon is required to print this material; use part number A62-R4300.
7 Options Available Below
Product Details & Documents
Use high-adhesive, self-laminating labels for tough conditions like high-humidity, small diameter wires, and newer wire jacketing materials. Bulk label rolls are ideal for continuous printing, reducing the frequency of label changeouts. Wraptor™ A6200 bulk label roll adapter required to utilize bulk label rolls.
- Permanent acrylic adhesive
- Matte finish
- Self-laminating, self-extinguishing material
- Abrasion-resistant, chemical-resistant, humidity-resistant, oil-resistant, water-resistant, and weather-resistant
- Service temperature: −40 to 158 °F
Dependable, long-term adhesion to small diameter wires. Labels will stay adhered in even the most humid environments. Strong adhesion to a variety of wire jacketing materials, including Teflon™, silicone, braided and traditional PVC. Labels feature a white printable area and a clear overlaminating area that protects the print. Labels are self-extinguishing and resist chemicals, humidity, oil, and water.
Use with Wraptor™ A6200 bulk label roll adapter and Wraptor™ A6200 wrap printer applicator and Wraptor™ A6200 bulk label roll adapter (required). A ribbon is required to print this material; use part number A62-R4300.
Product Family Options
Product Information
- DésignationMatièrePour l'utilisation avecWidthHauteurPackagingAvailabilityPrice
- B-427 self-laminating vinyl wire labels, 22 - 12 AWGSelf-laminating VinylWraptor™ A62000.5"0.75"6400/rollLog in for availabilityRoll of 1$958.63Supplier Number: 170984
- B-427 self-laminating vinyl wire labels, 12 - 6 AWGSelf-laminating VinylWraptor™ A62000.5"1.25"4000/rollLog in for availabilityRoll of 1$674.90Supplier Number: 170985
- B-427 self-laminating vinyl wire labels, 10 - 6 AWGSelf-laminating VinylWraptor™ A62000.5"1.5"3400/rollLog in for availabilityRoll of 1$743.69Supplier Number: 170986
- B-427 self-laminating vinyl wire labels, 22 - 12 AWGSelf-laminating VinylWraptor™ A62001"0.75"6400/rollLog in for availabilityRoll of 1$1,369.18Supplier Number: 170987
- B-427 self-laminating vinyl wire labels, 16 - 8 AWGSelf-laminating VinylWraptor™ A62001"1"5000/rollLog in for availabilityRoll of 1$1,078.98Supplier Number: 170988
- B-427 self-laminating vinyl wire labels, 12 - 6 AWGSelf-laminating VinylWraptor™ A62001"1.25"4000/rollLog in for availabilityRoll of 1$932.82Supplier Number: 170989
- B-427 self-laminating vinyl wire labels, 10 - 6 AWGSelf-laminating VinylWraptor™ A62001"1.5"3400/rollLog in for availabilityRoll of 1$891.98Supplier Number: 170990