Multivariate Concentric Square Field Tests | Avantor
Multivariate Concentric Square Field Tests
Supplier:   MAZE ENGINEERS
Multivariate Concentric Square Field Tests
Supplier:  MAZE ENGINEERS
Certificates
Multivariate Concentric Square Field Tests
Multivariate Concentric Square Field Tests
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Multivariate Concentric Square Field test (MCSF) is a novel behavior model for investigation of risk assessment, anxiety, and shelter seeking behavior in rodents.
  • Non-reflective acrylic surfaces
  • Easy to clean
The MCSF was originally described in the literature by Meyerson et al., 2006, for rats and later adopted by Ekmark-Lewen for mice. The behavioral model provides several areas for the animal to explore by free choice, including sheltered, open, and elevated areas.
The MCSF mouse model features a 72×72 cm open field area, with a 42×42 cm central chamber. The MCSF rat model features a 95×95 cm open field area, with a 55×55 cm central chamber.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Matière
    Availability
    Price
  • Multivariate concentric square field test, mouse
    White, black, gray, blue, clear
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $12,943.41
    $12,943.41
    Each
       $12,943.41
    Supplier Number: 6601
  • Multivariate concentric square field test, rat
    White, black, gray, blue, clear
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10,612.72
    $10,612.72
    Each
       $10,612.72
    Supplier Number: 6602