Stairway Tests
Supplier:   MAZE ENGINEERS
Stairway Tests
Supplier:  MAZE ENGINEERS
Certificates
Stairway Tests
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Stairway test apparatus is a sensitive test for subtle perturbations in motor function.
  • Acrylic frame and metal rods
  • Easy to clean, assemble, and disassemble
The Stairway test apparatus requires little pretraining and is a more sensitive test for subtle perturbations in motor function.
The Stairway test for mouse features 28 rungs; 2.5 cm in diameter, spaced evenly every 1.5 cm. The base of the triangle is 60 cm wide, and the length on both sides is 70 cm. The Stairway test for rat features 42 rungs; 4 cm in diameter, spaced evenly every 2 cm. The base of the triangle is 90 cm wide, and the length on both sides is 100 cm.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Availability
    Price
  • Stairway test, mouse
    Acrylic/metal
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,077.16
    $4,077.16
    Each
       $4,077.16
    Supplier Number: 5013
  • Stairway test, rat
    Acrylic/metal
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,292.89
    $4,292.89
    Each
       $4,292.89
    Supplier Number: 5028