Visual Burrow System | Avantor
Visual Burrow System
Supplier:   MAZE ENGINEERS
Visual Burrow System
Supplier:  MAZE ENGINEERS
Certificates
Visual Burrow System
Visual Burrow System
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Visual Burrow System (VBS) was developed by Bob Blanchard and Caroline Blanchard in the late 1980s. The VBS is commonly used in the evaluation of social behaviors.
  • Default black acrylic construction
  • Easy to clean
The Visual Burrow System (VBS) mimics the natural environment of rodents, enabling observation of natural social interactions that would have been otherwise impossible to observe in the wild. Each colony in the Visual Burrow System is housed in a rectangular acrylic chamber. Three chambers are positioned behind a barrier wall in the burrow area, with a wall separating it from the open area. These chambers are connected to and opened through the wall via clear Plexiglas tubes.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Longueur
    Largeur
    Hauteur
    Matière
    Availability
    Price
  • Visual burrow system, mouse
    Black
    86 cm
    61 cm
    26 cm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,714.40
    $6,714.40
    Each
       $6,714.40
    Supplier Number: 4909
  • Visual burrow system, rat
    Black
    129 cm
    92 cm
    39 cm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,450.11
    $6,450.11
    Each
       $6,450.11
    Supplier Number: 4910