Self-Administration Chambers | Avantor
Self-Administration Chambers
Supplier:   MAZE ENGINEERS
Self-Administration Chambers
Supplier:  MAZE ENGINEERS
Certificates
Self-Administration Chambers
Self-Administration Chambers
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The self-administration chamber is used to assess reward-seeking and addiction behaviors. This chamber offers everything needed for operant-led self-administration assessments associated with food restriction and intravenous drug administration.
  • Rodent chamber with two nose poke apertures or levers
  • Two cue lights in the apertures
  • One pellet dispenser, infusion pump, frequency tone, shock grid, and shocker (optional)
Data analysis which can be performed with the self-administration chamber include: number of infusions; number of active lever presses or nose-pokes; number of inactive lever presses or nose-pokes.
The chamber comes with a sound-attenuating cubicle with light or fan, a syringe pump system, a cage with two portlights and retractable levers, a shock floor grid, and two pellet receptacles. The option for an optic sensor to detect nose pokes can be added.
Ordering information: The Self-administration chamber requires software to function (see accessory option). Software package is designed for use with the Self administration apparatus as standalone software.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Largeur
    Hauteur
    Availability
    Price
  • Self-administration chamber; mouse
    18 cm
    18 cm
    20 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $17,020.52
    $17,020.52
    Each
       $17,020.52
    Supplier Number: 5841
  • Self-administration chamber; rat
    26 cm
    26 cm
    25 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $17,667.69
    $17,667.69
    Each
       $17,667.69
    Supplier Number: 5842