Puzzle Boxes
Puzzle Boxes
Supplier:   MAZE ENGINEERS
Puzzle Boxes
Supplier:  MAZE ENGINEERS
Certificates
Puzzle Boxes
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The puzzle box was described by Deacon and colleagues in 2011 as a variant of the work of Galsworthy et al., (2002, 2005).
  • Acrylic construction
  • Non-reflective surfaces
Subjects are placed in a brightly lit compartment and quickly develop a preference for a smaller, dark goal compartment due to light/dark motivation. Subjects are challenged with various interruptions of increasing difficulties, and are tasked to adopt solutions to each new problem.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Matière
    Availability
    Price
  • Puzzle box; mouse
    White, black, grey, blue, clear
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,656.50
    $3,656.50
    Each
       $3,656.50
    Supplier Number: 4703
  • Puzzle box; rat
    White, black, grey, blue, clear
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,872.24
    $3,872.24
    Each
       $3,872.24
    Supplier Number: 4704