Conditioned Place Preference (CPP), Spatial with Corner Inserts | Avantor
Conditioned Place Preference (CPP), Spatial with Corner Inserts
Supplier:   MAZE ENGINEERS
Conditioned Place Preference (CPP), Spatial with Corner Inserts
Supplier:  MAZE ENGINEERS
Certificates
Conditioned Place Preference (CPP), Spatial with Corner Inserts
Conditioned Place Preference (CPP), Spatial with Corner Inserts
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The conditioned place preference chamber is a paradigm widely used to explore the reinforcing effects of natural and stimuli. The spatial conditioned place preference chamber is a modification of the original CPP chamber that assesses the similar paradigms as the standard CPP chamber, but includes standing inserts for spatial place preference procedures.
  • Acrylic construction
  • Easy to clean
  • Odor-free
  • Customizable colors for walls, floors, and corners also available
In this variant, subjects are allowed to freely move between a compartment in which they were conditioned with either cues or neutral cues. The different cues provide visual reinforcement. This dual chamber place preference allows for biased and unbiased conditioned place preference testing. A removable door allows isolation into one compartment of the apparatus for the subject. Preference testing is then done by removing the door to allow the subject to freely explore between the two compartments.
The Spatial conditioned place preference chamber is a modification of the original CPP chamber that assesses the similar paradigms as the standard CPP chamber but includes standing inserts for spatial place preference procedures. Combinations of floor and cues are available including solid colors of black, grey, white, clear, and blue, and patterns such as dots, and horizontal/vertical/diagonal stripes.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Longueur
    Largeur
    Hauteur
    Matière
    Availability
    Price
  • Spatial conditioned place preference; mouse
    Grey/clear
    27 cm
    46 cm
    30 cm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,077.16
    $4,077.16
    Each
       $4,077.16
    Supplier Number: 4401
  • Conditioned place preference (CPP); mouse
    Grey/clear
    27 cm
    46 cm
    30 cm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,998.56
    $2,998.56
    Each
       $2,998.56
    Supplier Number: 4403
  • Real-time place preference chamber; mouse
    White, black, grey, blue, clear
    60 cm
    30 cm
    30 cm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,077.16
    $4,077.16
    Each
       $4,077.16
    Supplier Number: 4421
  • Spatial conditioned place preference; rat
    Grey/clear
    47 cm
    86 cm
    40 cm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,234.40
    $6,234.40
    Each
       $6,234.40
    Supplier Number: 4402
  • Conditioned place preference (CPP); rat
    Grey/clear
    47 cm
    86 cm
    40 cm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,940.06
    $4,940.06
    Each
       $4,940.06
    Supplier Number: 4404
  • Real-time place preference chamber; rat
    White, black, grey, blue, clear
    80 cm
    40 cm
    40 cm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,292.89
    $4,292.89
    Each
       $4,292.89
    Supplier Number: 4422