Elevated Zero Mazes
Supplier:   MAZE ENGINEERS
Elevated Zero Mazes
Supplier:  MAZE ENGINEERS
Certificates
Elevated Zero Mazes
Elevated Zero Mazes
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The elevated zero maze is an elevated, ring-shaped runway with the same amount of area devoted to adjacent open and closed quadrants, similar to the elevated plus maze to study anxiety behaviors.
  • Acrylic construction
  • Odor-free
  • Matted finish for non-reflective surfaces
  • Easy to clean
  • Easy assembly and disssembly with a removable base
The elevated zero maze is very similar to the elevated plus maze, but with the center region of the elevated plus maze removed. It has been pharmacologically validated with various anxiolytic drugs.
The design is created with an elevated circular platform with two walled, enclosed portions as well as two open portions, allowing for clear interpretations of murine behavior. Because mice tend to avoid open-spaced areas, the animals will preferentially spend more time in the enclosed walled area.
Product Family Options
Informations sur le produit
  • Désignation
    Ø
    Largeur
    Hauteur
    Availability
    Price
  • Elevated zero maze; mouse
    50 cm
    5 cm
    20 cm (wall), 61 cm (stand)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,077.16
    $4,077.16
    Each
       $4,077.16
    Supplier Number: 4001
  • Elevated zero maze; rat
    100 cm
    10 cm
    30 cm (wall), 61 cm (stand)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,940.06
    $4,940.06
    Each
       $4,940.06
    Supplier Number: 4002