Motorized Running Wheels | Avantor
Motorized Running Wheels
Supplier:   MAZE ENGINEERS
Motorized Running Wheels
Supplier:  MAZE ENGINEERS
Certificates
Motorized Running Wheels
Motorized Running Wheels
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Motorized Running Wheel (MRW) is used for exercise training in rodents.
  • Simple and easy to construct and clean
The Motorized Running Wheel (MRW) consists of a centrally motorized wheel which allows forced running at low and intermediate intensity levels. The motorized running wheel is constructed using clear acrylic, with a running track made of bars. On one side of the wheel, a quarter-circle serves as the entrance and exit for the subject. Two triangular acrylic columns support the running wheel frame above the surface. Eight infrared sensors are placed at 70° between 0 to 140° (effective area of exercise), using thick semi-circular, transparent acrylic sheets attached at the external sides of the two triangular columns. The wheel is centrally motorized and comes with associated software that allows control of training intensity (low and intermediate levels).
Ordering information: The motorized running wheel requires software to function (see accessory option). Software package is designed for use with the Motorized Running Wheel as a standalone software package.
Product Family Options
Informations sur le produit
  • Désignation
    Largeur
    Availability
    Price
  • Motorized running wheel, mouse
    6 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,234.40
    $6,234.40
    Each
       $6,234.40
    Supplier Number: 5022
  • Motorized running wheel, rat
    10 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,450.11
    $6,450.11
    Each
       $6,450.11
    Supplier Number: 5023