Tilt Ladders
Supplier:   MAZE ENGINEERS
Tilt Ladders
Supplier:  MAZE ENGINEERS
Certificates
Tilt Ladders
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The rodent tilt ladder is used in many other motor function experiments, as it is combined with a ladder-like grounding and moved to any angle needed to obtain a reward.
  • Acrylic frame and metal rods
  • Easy to clean, assemble, and disassemble
The rodent tilt ladder was originally described in the literature in 1989 in physiologic saphenous vein experiments, where rodents were kept in a tilt state.
The tilt ladder for mouse features 40 ladder rungs, with a bar length of 10 cm to accommodate 8 cm width of ladder. The tilt ladder for rat features 45 ladder rungs, with a bar length of 15 cm to accommodate 13 cm width of ladder. For both models, the bar diameter is 0.3 cm, and drilled holes in regularly spaced 1cm intervals allow for multiple patterns of bar insertion.
Product Family Options
Product Information
  • Désignation
    Longueur
    Matière
    Availability
    Price
  • Tilt ladder, mouse
    60 cm
    Acrylic/metal
    Log in for availability
    Log in for availability
    Each
    Each - 3 861,44 $
    3 861,44 $
    Each
       3 861,44 $
    Supplier Number: 5014
  • Tilt ladder, rat
    65 cm
    Acrylic/metal
    Log in for availability
    Log in for availability
    Each
    Each - 4 077,16 $
    4 077,16 $
    Each
       4 077,16 $
    Supplier Number: 5015