T-Maze
T-Maze
T-Maze
T-Maze
Similar to the Y-Maze, T-Maze is a behavioral test used to assess spatial learning and memory in rodents.
  • Acrylic construction
  • Easy assembly and disssembly with a removable base
  • Easy to clean
The T-Maze is a behavioral test used to assess spatial learning and memory in rodents. The T-Maze consists of a starting arm and two goal arms, arranged in the shape of a 'T'. The goal arms can be baited with food or other rewards. The subject's movements are then tracked, and different measures such as the number of arm entries, the time spent in each arm, and the number of errors made can be used to assess the subject's memory and spatial navigation abilities.
The T-Maze for mouse measures 66 cm across the T-Maze. The T-Maze for rat measures 110 cm across the T-Maze. The T-Maze with central partition for mouse measures 70 cm across the T-Maze, with a central partition option.
Product Family Options
  • Désignation
    Color
    Longueur
    Largeur
    Hauteur
    Matière
  • T-maze, mouse
    Blue, black, gray, white, or clear
    30 cm (stem), 30 cm (arm)
    6 cm
    20 cm (wall)
    Acrylic
    Each
    Each - $2,567.11
    $2,567.11
    Each
       $2,567.11
     : 3401
  • T-maze, rat
    Blue, black, gray, white, or clear
    50 cm (stem), 50 cm (arm)
    10 cm
    30 cm (wall)
    Acrylic
    Each
    Each - $3,861.44
    $3,861.44
    Each
       $3,861.44
     : 3402
  • T-maze with central partition, mouse
    Blue, black, gray, white, or clear
    30 cm (stem), 30 cm (arm)
    10 cm
    20 cm (wall)
    Acrylic
    Each
    Each - $2,782.82
    $2,782.82
    Each
       $2,782.82
     : 3403
Accessories
  •  
    Description
  • 76669-922
    Food wells, Ø 2×1 cm, with lid, acrylic
    Each - $215.74
  •  
    Description
  • 76669-924
    T-maze stand, mouse, stainless steel, height: 32 cm
    Each - $566.27
  •  
    Description
  • 76669-926
    T-maze stand, rat, stainless steel, height: 45 cm
    Each - $647.19