About this item
Manufactured specifically for laser printing, the series 215 biopsy cassettes come with separate lids and are available in 12 colors.
- Series 215 laser cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ laser printers
- They contain a special constituent chemical compound that carbonizes when exposed to the laser beam
- Comes in 12 different colors: white, yellow, blue, pink, green, orange, gray, aqua, lilac, beige, ochre and red with color-matched lids
- Taped stacks of 40 cassettes; 25 stacks per case for a total of 1000 cassettes and lids
- Series 215 Laser Cassette is not compatible with thermal tape style printers
Designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into the Epedia™ laser cassette hoppers for printing.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.