Cole-Parmer® Crucible Tongs and Rack
Fournisseur: SPEX
Certificats
À propos de cet article
Safely handle crucibles with tongs and rack.
- Compatible with graphite crucibles
- Nickel-chromium (nichrome) construction
Use crucible rack and tongs to safely handle graphite crucibles.
Frequently Bought Together