Erlenmeyer Flasks with Joint
Supplier: United Scientific Supplies
Jump to:
À propos de cet article
Erlenmeyer flasks that can use a glass stopper.
- Made from ASTM E-438 Type-1 Class A, Borosilicate 3.3 glass
- Flasks include ground joints manufactured per ASTM E-1404, Type 2, Class-1 requirement
- Flasks are printed in white color enamel
Borosilicate glass flasks are designed to provide the best combination of thermal shock resistance and physical strength. They feature 24/40 jointed sockets and graduations in durable white enamel.
6 Options Available Below
Product Details & Documents
Erlenmeyer flasks that can use a glass stopper.
- Made from ASTM E-438 Type-1 Class A, Borosilicate 3.3 glass
- Flasks include ground joints manufactured per ASTM E-1404, Type 2, Class-1 requirement
- Flasks are printed in white color enamel
Borosilicate glass flasks are designed to provide the best combination of thermal shock resistance and physical strength. They feature 24/40 jointed sockets and graduations in durable white enamel.
Product Family Options
Informations sur le produit
- CapacitéST Joint No.AvailabilityPrice
- 125 ml24/40Log in pour la disponibilitéLog in pour la disponibilitéPack of 2Pack of 2 - $46.42$46.42Prix promotionnel pour les nouveaux clientsPack of 2$46.42Supplier Number: UNJTERFL125
- 250 ml24/40Log in pour la disponibilitéLog in pour la disponibilitéPack of 2Pack of 2 - $54.21$54.21Prix promotionnel pour les nouveaux clientsPack of 2$54.21Supplier Number: UNJTERFL250
- 300 ml24/40Log in pour la disponibilitéLog in pour la disponibilitéPack of 2Pack of 2 - $55.88$55.88Prix promotionnel pour les nouveaux clientsPack of 2$55.88Supplier Number: UNJTERFL300
- 500 ml24/40Log in pour la disponibilitéLog in pour la disponibilitéPack of 2Pack of 2 - $75.24$75.24Prix promotionnel pour les nouveaux clientsPack of 2$75.24Supplier Number: UNJTERFL500
- Supplier Number: UNJTERFL1000
- Supplier Number: UNJTERFL2000