Cole-Parmer® Ceramic Grinding Cylinders
Fournisseur: SPEX
Certificats
À propos de cet article
Angle-cut ends help shear sample matrix for thorough extraction of analyte into the solvent.
- Constructed of durable ceramic
- Use with standard 50 ml centrifuge tubes
Ceramic grinding media are excellent for mixing samples for extraction methods such as pesticide residue analysis (QuEChERS) or extracting oil from fish and seafood. Typically two cylinders are used per sample vial.
Frequently Bought Together