ESD Cleanroom Frocks
À propos de cet article
ESD-safe integrated cleanroom wear designed for mobile users in controlled environments.
- TX4000 ESD Grid Fabric; 99% polyester/1% carbon
- Raglan sleeves with ESD knit cuffs for body contact
- Military collar
- Zipper closure with snaps for easy gowning
- Seams sewn with conductive threads and grounding tape for reliable panel-to-panel electrical continuity
- Reusable/launderable
- ISO cleanroom: ISO 6 (Class 1000)
- Meets standards required for ANSI/ESD S20.20
The Transforming Technologies' TX4000 ESD Cleanroom Garment System protects the controlled environment from the number one source of contamination and static found in cleanrooms - people. Electrostatic charge is an invisible contaminant that has the potential to degrade or destroy both product and equipment, as well as attract unwanted particles. Controlling these charges on mobile operators with the TX4000 Garment System throughout the controlled environment is essential to a productive manufacturing process.
Certifications: In compliance with IEST-RP-CC052.1, ANSI/ESD S20.20.
12 Options Available Below
Product Details & Documents
ESD-safe integrated cleanroom wear designed for mobile users in controlled environments.
- TX4000 ESD Grid Fabric; 99% polyester/1% carbon
- Raglan sleeves with ESD knit cuffs for body contact
- Military collar
- Zipper closure with snaps for easy gowning
- Seams sewn with conductive threads and grounding tape for reliable panel-to-panel electrical continuity
- Reusable/launderable
- ISO cleanroom: ISO 6 (Class 1000)
- Meets standards required for ANSI/ESD S20.20
The Transforming Technologies' TX4000 ESD Cleanroom Garment System protects the controlled environment from the number one source of contamination and static found in cleanrooms - people. Electrostatic charge is an invisible contaminant that has the potential to degrade or destroy both product and equipment, as well as attract unwanted particles. Controlling these charges on mobile operators with the TX4000 Garment System throughout the controlled environment is essential to a productive manufacturing process.
Certifications: In compliance with IEST-RP-CC052.1, ANSI/ESD S20.20.
Specifications for Product Family
Spécifications
- Catalog No:
- 77008-130
- 77008-178
- 77008-092
- 77008-140
- 77008-192
- 77008-098
- 77008-166
- 77008-142
- 77008-102
- 77008-196
- 77008-144
- 77008-116
- 77008-204
- 77008-088
- 77008-132
- 77008-198
- 77008-114
- 77008-148
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Fabric:
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
Spécifications
Product Family Options
Informations sur le produit
- ColorTaille de confectionAvailabilityPrice
- NavyXSDiscontinued-Supplier Number: TX40FRNB01Spécifications:Marketsource
More
Specifications
Cat. No.77008-130Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbonMarketsource - Supplier Number: TX40FRNB02Spécifications:
More
Specifications
Cat. No.77008-178Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbon - Supplier Number: TX40FRNB03Spécifications:
More
Specifications
Cat. No.77008-092Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbon - Supplier Number: TX40FRNB04Spécifications:
More
Specifications
Cat. No.77008-140Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbon - Supplier Number: TX40FRNB05Spécifications:
More
Specifications
Cat. No.77008-192Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbon - Supplier Number: TX40FRNB06Spécifications:
More
Specifications
Cat. No.77008-098Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbon - Supplier Number: TX40FRNB07Spécifications:
More
Specifications
Cat. No.77008-166Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbon - Navy4XLDiscontinued-Supplier Number: TX40FRNB08Spécifications:Marketsource
More
Specifications
Cat. No.77008-142Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbonMarketsource - Navy5XLDiscontinued-Supplier Number: TX40FRNB09Spécifications:Marketsource
More
Specifications
Cat. No.77008-102Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbonMarketsource - Supplier Number: TX40FRWH01Spécifications:
More
Specifications
Cat. No.77008-196Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbon - Supplier Number: TX40FRWH02Spécifications:
More
Specifications
Cat. No.77008-144Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbon - Supplier Number: TX40FRWH03Spécifications:
More
Specifications
Cat. No.77008-116Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbon - Supplier Number: TX40FRWH04Spécifications:
More
Specifications
Cat. No.77008-204Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbon - Supplier Number: TX40FRWH05Spécifications:
More
Specifications
Cat. No.77008-088Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbon - Supplier Number: TX40FRWH06Spécifications:
More
Specifications
Cat. No.77008-132Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbon - White3XLDiscontinued-Supplier Number: TX40FRWH07Spécifications:Marketsource
More
Specifications
Cat. No.77008-198Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbonMarketsource - White4XLDiscontinued-Supplier Number: TX40FRWH08Spécifications:Marketsource
More
Specifications
Cat. No.77008-114Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbonMarketsource - White5XLDiscontinued-Supplier Number: TX40FRWH09Spécifications:Marketsource
More
Specifications
Cat. No.77008-148Garment applicationCleanroomRisk areaESDFabric99% polyester/1% carbonMarketsource