Anti-phospho-Ser/Thr ATM / ATR Substrate Rabbit Polyclonal Antibody
Fournisseur: ANTIBODIES.COM LLC
Certificats
À propos de cet article
Rabbit polyclonal antibody to phospho-Ser/Thr ATM / ATR Substrate for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: phospho-Ser/Thr ATM / ATR Substrate
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 77208-304
- Antigen symbol:
- phospho-Ser/Thr ATM / ATR Substrate
- Antigen name:
- phospho-Ser/Thr ATM / ATR Substrate
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence containing phosphorylated S & T.
- Form:
- Liquid
- Molecular weight:
- 38 - 68 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
Spécifications
Frequently Bought Together