Anti-NME2 Mouse Monoclonal Antibody [clone: NME2/6433]
77177-708
- Form:Liquid
- Gene ID:UniprotID# P22392
- Antigen synonyms:epididymis secretory sperm binding protein Li 155an|C-myc purine-binding transcription factor PUF|HEL-S-155an|Histidine protein kinase NDKB|C myc transcription factor|NDKB_HUMAN|NDK B|MGC111212|MGC2212
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:17 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human NME2 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-708
- Packaging:Plastic vial
Mouse monoclonal [NME2/6433] antibody to NME2 for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: NME2
Clonality: Monoclonal
Clone: NME2/6433
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human