Anti-Carbonic Anhydrase 8/CA8 Mouse Monoclonal Antibody [clone: CA8/6814]
Catalogue # 77176-002
Fournisseur: ANTIBODIES.COM LLC
Spécifications
- Form:Liquid
- Gene ID:UniprotID# P35219
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:32.97 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CA8 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77176-002
- Packaging:Plastic vial
Spécifications
À propos de cet article
Mouse monoclonal [CA8/6814] antibody to Carbonic Anhydrase 8/CA8 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Carbonic Anhydrase 8/CA8
Clonality: Monoclonal
Clone: CA8/6814
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human