Anti-EF-Ts Rabbit Monoclonal Antibody [clone: ARC2918]
Fournisseur: Avantor
Certificats
À propos de cet article
Rabbit monoclonal [ARC2918] antibody to EF-Ts for WB, ICC/IF and IP with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ICC/IF, IP
- Recommended Dilutions: WB: 1:100 to 1:500, ICC/IF: 1:50 to 1:200, IP: 1:100 to 1:500
Type: Primary
Antigen: EF-Ts
Clonality: Monoclonal
Clone: ARC2918
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Frequently Bought Together