Anti-DPM3 Goat Polyclonal Antibody
Catalogue # 77069-404
Fournisseur: ANTIBODIES.COM LLC
Spécifications
- Antibody type:Primary
- Antigen name:CDG1O
- Antigen symbol:DPM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Dolichyl Phosphate beta D Mannosyltransferase subunit 3|Mannose P Dolichol Synthase subunit 3|MPD synthase subunit 3|Dolichyl phosphate mannosyltransferase polypeptide 3|DPM synthase complex subunit 3|DPM3|Dolichol Phosphate Mannose Synthase subunit 3|Dolichol phosphate mannosyltransferase subunit 3|Prostin 1
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:HDCEDAARELQSQ
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human DPM3 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77069-404
- Packaging:Plastic vial
Spécifications
À propos de cet article
Goat polyclonal antibody to DPM3 for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: DPM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human