Anti-Erythrocyte Specific Mouse Monoclonal Antibody [clone: SFL23.6]
Catalogue # 76950-036
Fournisseur: Antibodies.com
Spécifications
- Antibody type:Primary
- Antigen symbol:Erythrocyte Specific
- Clonality:Monoclonal
- Clone:SFL23.6
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Hepatocytes from a 20-22 week-old human fetus
- Purification:Protein A/G chromatography
- Cat. no.:76950-036
- Packaging:Plastic vial
Spécifications
À propos de cet article
Mouse monoclonal [SFL23.6] antibody to Erythrocyte Specific for Flow Cytometry and IF with samples derived from Human.
- Validated Applications: Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: Erythrocyte Specific
Clonality: Monoclonal
Clone: SFL23.6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human