ContactUsLinkComponent
Lab Armor® Bead Baths, 20L, 230 V, Shel Lab, Sheldon
Fournisseur:  Sheldon Manufacturing
Certificats
undefined
Lab Armor® Bead Baths, 20L, 230 V, Shel Lab, Sheldon
Fournisseur:  Sheldon Manufacturing
Spécifications
  • Catalog No:
  • 89409-236
  • Dimensions:
  • 15.2 x 30.5 x 43.2 cm (6 x 12 x 17"), tub
  • Exterior dimensions:
  • 21.6 x 40.4 x 59 cm (8.5 x 15.9 x 23.25")