Anti-RAIDD Rabbit Monoclonal Antibody [Clone: ARC1771]
Fournisseur: ANTIBODIES.COM LLC
Certificats
À propos de cet article
Rabbit monoclonal [ARC1771] antibody to RAIDD for WB and IHC with samples derived from Human and Rat.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200
Type: Primary
Antigen: RAIDD
Clonality: Monoclonal
Clone: ARC1771
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Frequently Bought Together