Anti-Tyrosyl tRNA synthetase Rabbit Polyclonal Antibody
77052-974
- Antibody type:Primary
- Antigen name:CMTDIC
- Antigen symbol:Tyrosyl tRNA synthetase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P54577
- Antigen synonyms:TyrRS|yars|Tyrosyl-tRNA synthetase, cytoplasmic|YRS|Tyrosine tRNA ligase, cytoplasmic|Tyrosyl tRNA synthetase|Tyrosyl--tRNA ligase|Tyrosine tRNA ligase 1, cytoplasmic|SYYC_HUMAN
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:59 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from the C-terminal region of human YARS (amino acids 451-500)
- Tested applications:IHC
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Cat. no.:77052-974
- Packaging:Plastic vial
Rabbit polyclonal antibody to YARS for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:2000, IHC-P: 1:50-1:300, ELISA: 1:10000
Type: Primary
Antigen: Tyrosyl tRNA synthetase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat