Anti-GPCR GPR31 Rabbit Polyclonal Antibody
77079-746
- Antibody type:Primary
- Antigen name:12-(S)-HETE acid receptor
- Antigen symbol:GPCR GPR31
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O00270
- Antigen synonyms:bA517H2.2 (G protein coupled receptor 31)|GPCR GPR31|12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid receptor|G protein coupled receptor 31|12-(S)-HYDROXYEICOSATETRAENOIC ACID RECEPTOR|12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid (HETE) receptor|HETER1|12-HETER|OTTHUMP00000017616
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:35 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human GPR31 (amino acids 181-230)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77079-746
- Packaging:Plastic vial
Rabbit polyclonal antibody to GPR31 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000
Type: Primary
Antigen: GPCR GPR31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human