Anti-GPCR GPR103 Rabbit Polyclonal Antibody
Catalogue # 77080-074
Fournisseur: ANTIBODIES.COM LLC
Spécifications
- Antibody type:Primary
- Antigen name:AQ27
- Antigen symbol:GPCR GPR103
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q96P65
- Antigen synonyms:Orexigenic neuropeptide QRFP receptor|QRFPR|QRFPR_HUMAN|SP9155|G-protein coupled receptor 103|G protein-coupled receptor 103|GPCR GPR103|Pyroglutamylated RFamide peptide receptor
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:49 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human GPR103 (amino acids 351-400)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77080-074
- Packaging:Plastic vial
Spécifications
À propos de cet article
Rabbit polyclonal antibody to GPR103 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: GPCR GPR103
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human