Life Functions of Paramecia Microslides
Fournisseur: SUPERTEK SCIENTIFIC LLC
Certificats
À propos de cet article
Explore the daily functions of paramecium.
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects It on both sides
- Set of 15 Includes detailed lesson pan on the topic under study
- Durable, inexpensive teaching resource
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery: Set Includes: 1. Paramecium with bacteria (100×)/Paramecia with air bubble (40×) 2. Ingestion (200×)/Digestion (100×) 3. Paramecium dividing (100×) 4. Paramecium (200×) 5. Contractile vacuoles (200×) 6. Paramecium nuclei (200×)/Conjugation (100×) 7. Paramamecium bending (100×) 8. Trichocysts (100×).
Frequently Bought Together