Cleavage Microslides
Learn about this important step of cell divison.
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects It on both sides
- Set of 15 Includes detailed lesson pan on the topic under study
- Durable, inexpensive teaching resource
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
: Viewers are available separately.
: Set Includes: 1. One-cell stage (320×) 2. Two-cell stage (320×) 3. Four-cell stage (320×) 4. Eight-cell stage (320×) 5. A mass of cells (320×) 6. The Blastula (320×) 7. Early Gastrula (160×) 8. Late Gastrula (240×).
Frequently Bought Together