ContactUsLinkComponent
ZymoPURE™ 96 Plasmid Miniprep Kits
ZymoPURE™ 96 Plasmid Miniprep Kits
Supplier:  Zymo Research
Certificates
undefined
ZymoPURE™ 96 Plasmid Miniprep Kits
Supplier:  Zymo Research
Specifications
  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid (endotoxin-free)
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid