FKBP12 Peptide
Vial, 0.05 mg
77128-102
: Prosci
: 7789P
- Type de conditionnement:Vial
- Taille:0.05 mg
- Cat. No.:77128-102
- Protein/peptide type:Synthetic
- Protein/peptide name:FKBP1A
- Storage conditions:Store FKBP12 Peptide at −20 °C, stable for one year.
- Gene ID:2280
- Formulation:Liquid. PBS pH 7.2 (10 mM NaH₂PO₄, 10 mM Na₂HPO₄, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide.
- Protein synonyms:PKC12|FKBP-12|FKBP1|PPIASE|PKCI2|FKBP-1A|PPIase FKBP1A|Peptidyl-prolyl cis-trans isomerase FKBP1A|FKBP12|12 kDa FK506-binding protein
- Conjugation:Unconjugated
- Concentration:200 µg/ml
- Tested applications:Blocking
- Species:Human
- Shipping temperature:20
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



