Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
SMUG1 Peptide

SMUG1 Peptide

Vial, 0.1 mg
Catalog # 77124-478
Supplier:   Prosci
Supplier #:  46-390P
SMUG1 Peptide
Catalog # 77124-478
Supplier:  Prosci
CAS Number:  
SMUG1 Peptide

Vial, 0.1 mg

CAS Number:  
Price
$477.16
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Type de conditionnement:
    Vial
  • Taille:
    0.1 mg
  • Cat. No.:
    77124-478
  • Protein/peptide type:
    Synthetic
  • Protein/peptide name:
    SMUG1
  • Storage conditions:
    Store at −20 °C. When reconstituted minimize freezing and thawing.
  • Gene ID:
    23583
  • Formulation:
    Lyophilized. SMUG1 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
  • Protein synonyms:
    HMUDG|UNG3|Single-strand selective monofunctional uracil DNA glycosylase|FDG
  • Conjugation:
    Unconjugated
  • Tested applications:
    Blocking
  • Species:
    Human
  • Shipping temperature:
    RT

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.