Mouse Recombinant Complement C5 Protein (from HEK293 Cells)
Vial, 0.1 mg
Catalog # 77120-658
Supplier: Prosci
Fournisseur #: 11-357
Mouse Recombinant Complement C5 Protein (from HEK293 Cells)
Catalog # 77120-658
Supplier: Prosci
CAS Number:
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Spécifications
Spécifications
- Type de conditionnement:Vial
- Taille:0.1 mg
- Protein/peptide name:Complement C5
- Protein synonyms:CPAMD4|Complement C5|C5
- Purity:>95% as determined by SDS-PAGE
- Sequence:Gln 19 - Glu 1680
- Source:HEK293 cells
- Molecular weight:73.5 kDa (β chain) and 114.6 kDa (α chain)
- Shipping temperature:RT
- Cat. No.:77120-658
- Protein/peptide type:Recombinant
- Storage conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Formulation:Lyophilized. PBS, pH 7.4.
- Conjugation:Unconjugated
- Tested applications:ELISA, WB
- Species:Mouse
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



