Anti-GHRL Rabbit Polyclonal Antibody
50 µl, Vial
Catalog # 76763-322
Supplier: Prosci
Supplier #: 14-959
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type: Primary
Antigen: GHRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Taille:50 µl
- Packaging:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Modification:None
- Purification:Affinity purification
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Concentration:Batch dependent
- Shipping temperature:20
- Antigen name:Ghrelin and obestatin prepropeptide
- Cat. No.:76763-322
- Reactivity:Human
- Gene ID:51738
- Antigen synonyms:ghrelin/obestatin prepropeptide|motilin-related peptide|preproghrelin|In2c-preproghrelin|growth hormone-releasing peptide|ghrelin|growth hormone secretagogue receptor ligand|MTLRP|appetite-regulating hormone|ghrelin/obestatin preprohormone|prepro-appetite regulatory hormone
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-117 of human GHRL (NP_057446.1).
- Conjugation:Unconjugated
- Antigen symbol:GHRL
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3



