ERV3 Peptide
Vial, 0.1 mg
77122-008
: Prosci
: 42-697P
- Type de conditionnement:Vial
- Taille:0.1 mg
- Cat. No.:77122-008
- Protein/peptide type:Synthetic
- Protein/peptide name:ERV3-1
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:2086
- Formulation:Lyophilized. ERV3 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Protein synonyms:ERV3|HERV-R|HERVR|ERV-R|ERV-3 envelope protein|ERVR|ERV3 envelope protein|envR
- Conjugation:Unconjugated
- Tested applications:Blocking
- Shipping temperature:RT



