10-Level Conductivity Indicators | Avantor
10-Level Conductivity Indicators
Supplier:   GSC International
10-Level Conductivity Indicators
Supplier:  GSC International
Certificates
10-Level Conductivity Indicators
10-Level Conductivity Indicators
2 Options Available Below
Product Details & Documents
A very simple and visual demonstration of the level of conductivity of almost any solution.
  • Simple to use
  • Battery included and can be replaced as needed
  • LED Indicator light are a quick visual confirmation of the level of conductivity
  • Two pack sizes available
The 10-Level conductivity indicator can be used for quick comparisons of conductivity in solutions including, but not limited to, tap water, vinegar, baking soda solution, or varying concentrations of salt water. This device is very simple to use, just submerge both brass prongs in a solution hold the button on the side, and wait for the bar of LED's to display your results. Instruction are included.
Caution: Prop 65 Warning Label is listed on the package for lead and BPA.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • 10-level conductivity indicator
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $70.95
    $70.95
    Each
       $70.95
    Supplier Number: CNDTEST
    Spécifications:
    More
    Specifications
    Cat. No.
    470352-982
  • 10-level conductivity indicator, 10 units per case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $675.00
    $675.00
    Pack of 10
       $675.00
    Supplier Number: CNDTEST-10
    Spécifications:
    More
    Specifications
    Cat. No.
    470352-976