ContactUsLinkComponent
Eisco Latex Dropper Bulbs
Catalogue # 470344-042
Fournisseur:  Wards
undefined
Eisco Latex Dropper Bulbs
Catalogue # 470344-042
Fournisseur:  Wards
Supplier Number:  CH0232DPK72
Spécifications
  • Ø:
    0.5"
  • Capacity:
    2 ml
  • Length:
    2.2"
  • Cat. no.:
    470344-042
Spécifications
À propos de cet article
Bulbs made from high-quality latex fit pipettes and medicine droppers measuring 6 to 8 mm in diameter.
  • Pack of 72 dropper bulbs made of high quality latex rubber
  • Excellent for science classrooms and professional laboratories
Latex dropper bulbs fit glass pipettes.