Alpha Carbin Program for Pipette Tip Boxes
Product CA78646-246 is a replacement for 76635-494 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Tired of plastic waste in the lab? So are we. With Polycarbin's Alpha Carbin program pipette tip boxes (from any vendor) can be transformed into the next generation of lab products. Track the progress and restock with low-carbon products to become a Closed-Loop lab.
- Accepts all brands of pipette tip boxes
- Prepaid return labels and carbon neutral shipping
- Personalized dashboard showing impact data
Today the vast majority of lab plastics are either burned or buried, perpetuating the 'take-make-waste' model of innovation. Polycarbin uses lab plastics as the key input in our manufacturing process, turning them into the next generation of lab products.
Polycarbin's Alpha Carbin program is a mailback service for any brand of pipette tip box. Simply fill the Carbin with empty pipette tip boxes. Once full, use the prepaid return labels to ship them back to Polycarbin. From here, users can track their efforts with Polycarbin's data platform, the Carbin Counter.
: Alpha Carbins should come with prepaid return labels affixed to each box. If Carbins do not have them upon arrival, please contact your Avantor representative.
: Alpha Carbins come in packs of five. The external package has dimensions of 27L×25W×10"H.
: The Alpha Carbin program is exclusively for clean, empty pipette tip boxes and no other plastics.
Specifications
- Capacité:Approx. 100 tip boxes
- Désignation:Alpha carbin mailback program for pipette tip boxes
- Dimensions:12×12×26"
- Matière:Cardboard
- Taille:16 gal.
- Cat. No.:76635-494
- Environmental certification:This environmental impact of this product was evaulated by a third party, panel reviewed LCA to determine recycled content, carbon dioxide emission eq. reduction, water conservation, and crude oil conservation.
- Environmentally Preferable:Waste Reducing|Product Transparency and Disclosure