ContactUsLinkComponent
Inverted Microscope with FOV20 and S80 Basic Stand Outfit, DMi1, Leica Microsystems
Inverted Microscope with FOV20 and S80 Basic Stand Outfit, DMi1, Leica Microsystems
Catalog # CA76538-910
Supplier:  Leica Microsystems
undefined
Inverted Microscope with FOV20 and S80 Basic Stand Outfit, DMi1, Leica Microsystems
Catalog # CA76538-910
Supplier:  Leica Microsystems
Supplier Number:  11526231
Specifications
  • Description:
    Inverted tissue culture microscope with 20 mm eyepieces, 80 mm WD condenser
  • Cat. no.:
    CA76538-910
Specifications
About this item
Microscope for basic cell culture applications and flasks.
  • 'Two eyepieces (10×/20 eyepiece with eyeguard and 10×/20 focusing eyepiece with eyeguard)
  • S80 condenser for flasks
  • 4 position slider for S80 condenser (BF, Ph0, Ph1, Ph2)
  • 3 objectives (HI PLAN 4×/0.10 PH0, HI PLAN I 10×/0.22 PH1, HI PLAN I 20×/0.30 PH1)
  • Dust cover
Leica DMi1 FOV20 basic stand (FOV20, LED illumination, exchangeable eyepieces, fixed stage, power cable).