ContactUsLinkComponent
Upright Microscope with FITC and TRITC Filter Fluorescence Bundle, BA410E
  CA77147-438
 :  Motic
undefined
Upright Microscope with FITC and TRITC Filter Fluorescence Bundle, BA410E
  CA77147-438
 :  Motic
 :  1100100404312
  • Model:
    Model BA410E
  • Description:
    Model BA410E with FITC and TRITC filter fluorescence bundle
  • Cat. no.:
    CA77147-438
  • Focal Length:
    Model BA410E with FITC and TRITC filter fluorescence bundle
This new fluorescence bundle includes BA410E 3WLED trinocular sextuple microscope (light split 80/20), complete EPI-fluorescence attachment, EGFP/FITC/Cy2/AlexaFluor 488 and TRITC/Cy3/TagRFP/AlexaFluor 546. Universities and laboratories with applications in hematology, cytology and histology will look for this flexible microscope platform with a superb performance in optics and mechanics.
  • Display maximum functionality and ergonomics in all its mechanical aspects
  • Elegant and robust, the microscope stand follows all needs for an intense daily use
  • Eyepiece tubes guarantee a fatigue-free work in terms of viewing angle and improved interpupillary distance
  • The 'butterfly' movement may increase the viewing height by 60 mm for individual positioning
  • CCIS© Infinity concept presents this apochromatic level of performance in a complete set of optional bright field objectives, compatible to all BA series models of upright microscopes
  • Highest quality demands from pathology, histology and hematology can now be satisfied for a reliable diagnosis in routine and research work of hospitals and other biomedical labs
  • Based on an encoded 6-fold nosepiece which memorizes the light intensity setting for each nosepiece position
  • The system replicates this intensity setting once the respective position is swung in again; any new setting is taken as an overwriting of the former setup
Create a digital environment with Moticam: digitize your lab with our Moticam solutions. Look at your samples in your computer, your tablet or smartphone, a TV screen or a projector and view, capture, edit, measure, make reports or share your work with ease with anyone in just a few clicks. Our software solutions included in the packages show a user-friendly interface, making data sharing possible in a collaborative environment, encouraging teamwork and collective learning experiences.