ContactUsLinkComponent
Upright Microscope with FITC Filter Fluorescence Bundle, Panthera C2
  CA77147-440
 :  Motic
undefined
Upright Microscope with FITC Filter Fluorescence Bundle, Panthera C2
  CA77147-440
 :  Motic
 :  1100104601331
  • Model:
    Panthera C2
  • Description:
    Panthera C2 with FITC filter fluorescence bundle
  • Cat. no.:
    CA77147-440
  • Focal Length:
    Panthera C2 with FITC filter fluorescence bundle
This new fluorescence bundle includes a Panthera C2 trinocular 50:50 microscope and a FITC filter (fluorescence accessory). If it is about stained sections with cover glass protection, about native smears, fine needle biopsies, blood smears which wanted to be treated with or without cover glass: Panthera C2 is your solution.
  • Bright field is the standard, phase contrast, pol contrast, objectives with/without cover glass correction, optional LED fluorescence modules with variable filter combinations
  • The new Plan UC Achromatic objectives, from 2× to 100× magnification range, show significantly improved image contrast for your individual application
  • The Panthera C2 offers flexibility in light sources
  • A 30 W Halogen bulb can be replaced by a 3 W LED of different color temperatures
  • The universal turret condenser with all phase rings and a darkfield stop; the combination of polarizer/analyzer for birefringent samples
Create a digital environment with Moticam: digitize your lab with our Moticam solutions. Look at your samples in your computer, your tablet or smartphone, a TV screen or a projector and view, capture, edit, measure, make reports or share your work with ease with anyone in just a few clicks. Our software solutions included in the packages show a user-friendly interface, making data sharing possible in a collaborative environment, encouraging teamwork and collective learning experiences.