Quick-DNA/RNA™ Miniprep Plus Kits
À propos de cet article
The Quick-DNA/RNA MiniPrep Plus kit combines Quick-DNA/RNA technology with the addition of DNA/RNA Shield, a preservation and lysis technology, and Proteinase K to enable easy, reliable, and rapid isolation from any biological sample including cells, solid tissue, and whole blood. The procedure uses Zymo-Spin column technology that results in high-quality genomic DNA and total RNA that is ready for any downstream application including reverse transcription, microarray, sequencing, etc.
2 Options Available Below
Product Details & Documents
The Quick-DNA/RNA MiniPrep Plus kit combines Quick-DNA/RNA technology with the addition of DNA/RNA Shield, a preservation and lysis technology, and Proteinase K to enable easy, reliable, and rapid isolation from any biological sample including cells, solid tissue, and whole blood. The procedure uses Zymo-Spin column technology that results in high-quality genomic DNA and total RNA that is ready for any downstream application including reverse transcription, microarray, sequencing, etc.
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- 77001-854
- 76358-832
- Binding capacity:
- 100 µg
- 100 µg
- Yield:
- 100 µg DNA and RNA (binding capacity), ≥50 µl (elution volume)
- 100 µg DNA and RNA (binding capacity), ≥50 µl (elution volume)
Spécifications
Product Family Options
Informations sur le produit
- DésignationDetailsTailleAvailabilityPrice
- Quick-DNA/RNA™ Miniprep Plusfor Cells, any tissues, blood, biological fluids50 PrepsEach$765.76Kit of 1$765.76Supplier Number: D7003L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.77001-854Binding capacity100 µgYield100 µg DNA and RNA (binding capacity), ≥50 µl (elution volume) - Quick-DNA/RNA™ Miniprep Plusfor Cells, any tissues, blood, biological fluids10 PrepsLog in pour la disponibilitéEach$186.65Supplier Number: D7003TSpécifications:
More
Specifications
Cat. No.76358-832Binding capacity100 µgYield100 µg DNA and RNA (binding capacity), ≥50 µl (elution volume)