Eisco Triangular Retort Bases | Avantor
Eisco Triangular Retort Bases
Supplier:   Wards
Eisco Triangular Retort Bases
Supplier:  Wards
Certificates
Eisco Triangular Retort Bases
Eisco Triangular Retort Bases
3 Options Available Below
Product Details & Documents
Cast iron Triangular Retort Stand with center-tapped hole for maximum stability in minimum footprint.
  • Three-point retort bases made of chemical-resistant cast iron with a black coating
  • Offers maximum stability in a minimal footprint
  • Sturdy construction provides superb support for experimental setups
  • Features a centered-tapped hole for a 10×1.5 mm threaded rod (not included)
Triangular 3-Point Retort Stands are made of chemically-resistant cast iron with a black finish. Features a centered tapped hole suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for both science classrooms and professional laboratories.
Product Family Options
Informations sur le produit
  • Longueur
    Weight
    Availability
    Price
  • 4"
    1.75 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $45.40
    $45.40
    Each
       $45.40
    Supplier Number: CH0648A
  • 5.25"
    2.2 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $49.00
    $49.00
    Each
       $49.00
    Supplier Number: CH0648B
  • 6.3"
    3.3 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $60.05
    $60.05
    Each
       $60.05
    Supplier Number: CH0648C
Accessories
Informations sur le produit
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Steel support rod
    Steel support rod
    470345-310
    12 mm
    508 mm (20")
    Log in pour la disponibilité
    Each - $17.00
Informations sur le produit
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Aluminum support rod
    Aluminum support rod
    470343-698
    12 mm
    609 mm (24")
    Log in pour la disponibilité
    Each - $19.10
Informations sur le produit
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Steel support rod
    Steel support rod
    470345-166
    12 mm
    762 mm (30")
    Log in pour la disponibilité
    Each - $18.40
Informations sur le produit
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Stainless steel support rod
    Stainless steel support rod
    470344-578
    12 mm
    762 mm (30")
    Log in pour la disponibilité
    Each - $28.95
Informations sur le produit
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Aluminum support rod
    Aluminum support rod
    470345-558
    12 mm
    508 mm (20")
    Log in pour la disponibilité
    Each - $17.05
Informations sur le produit
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Aluminum support rod
    Aluminum support rod
    470345-052
    12 mm
    762 mm (30")
    Log in pour la disponibilité
    Each - $25.40
Informations sur le produit
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Stainless steel support rod
    Stainless steel support rod
    470344-218
    12 mm
    609 mm (24")
    Log in pour la disponibilité
    Each - $24.25
Informations sur le produit
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Stainless steel support rod
    Stainless steel support rod
    470345-364
    12 mm
    508 mm (20")
    Log in pour la disponibilité
    Each - $22.65
Informations sur le produit
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Steel support rod
    Steel support rod
    470345-394
    12 mm
    609 mm (24")
    Log in pour la disponibilité
    Each - $22.65
Informations sur le produit
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Stainless steel support rod
    Stainless steel support rod
    470345-774
    12 mm
    1016 mm (40")
    Log in pour la disponibilité
    Each - $38.25
Informations sur le produit
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Aluminum support rod
    Aluminum support rod
    470345-328
    12 mm
    1016 mm (40")
    Log in pour la disponibilité
    Each - $31.35
Informations sur le produit
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Stainless steel support rod
    Stainless steel support rod
    470344-962
    12 mm
    901 mm (35.5")
    Log in pour la disponibilité
    Each - $38.25