Eisco Eudiometer Tubes
 :   Wards
Eisco Eudiometer Tubes
 :  Wards
Eisco Eudiometer Tubes
Eisco Eudiometer Tubes
Eudiometer tubes with platinum elecrodes for measuring gas volume changes during chemical reactions.
  • Made of mechanically strong and chemical resistant Borosilicate 3.3 glass
  • Contains two platinum electrodes on the sealed end of tube
  • Easy-to-read white graduations
Eudiometer tubes made of high quality Borosilicate glass. Useful for measuring volume of gas produced or consumed in a chemical reaction.
Product Family Options
  • Capacité
    Désignation
    Matière
  • 50 ml
    Eudiometer tubes, 0.1 ml graduations
    Borosilicate glass 3.3
    Each
    Each - $66.25
    $66.25
    Each
       $66.25
     : CH0499
  • 100 ml
    Eudiometer tubes, 0.2 ml graduations
    Borosilicate glass 3.3
    Each
    Each - $67.30
    $67.30
    Each
       $67.30
     : CH0499B